missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is used in artificial photosynthesis. It is used as a magnetic semiconductor. It is also employed in supercapacitors and lithium-ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 900°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN3077 |
Merck Index | 14,2429 |
Solubility Information | Insoluble in water.Soluble in acids and alkalies. |
Formula Weight | 240.8 |
Percent Purity | 99.9985% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Product Identifier
- Cobalt(II,III) oxide
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Cobalt oxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction