missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11378597
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15238-00-3
MDL Number MFCD00016030
Synonym cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis

Specifications

Boiling Point 570°C (vac.)
Melting Point 515°C (vac.)
Color White
Physical Form Beads
Quantity 5 g
Merck Index 14,2443
Formula Weight 312.74
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 5.584 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.