missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) nitrate hexahydrate, 97.7% min, Thermo Scientific Chemicals

Product Code. 11371109
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,2444
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 291.04 (182.94 Anhydrous)
Percent Purity ≤97.7%
Odor Odorless
Refractive Index 1.52
Density 1.87 g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'