missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™

Product Code. 15602290
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C
Color Red
Quantity 100 g
Linear Formula Co(NO3)2 · 6H2O
UN Number UN1477
IUPAC Name cobalt(2+);dinitrate;hexahydrate
Formula Weight 291.03g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals or chips
Density 1.87g/cm3
Chemical Name or Material Cobalt(Ii) Nitrate Hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'