missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate dihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

114.00 € - 483.00 €

Chemical Identifiers

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.981
MDL Number MFCD00050793
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name cobalt(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Co+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013012.09
10 g
114.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11372108
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013012.18
50 g
483.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5965-38-8
182.981
MWHSMSAKVHVSAS-UHFFFAOYSA-L
131881712
C(=O)(C(=O)[O-])[O-].O.O.[Co+2]
C2H4CoO6
MFCD00050793
cobaltousoxalate,cobaltoxalate
cobalt(2+);oxalate;dihydrate

Specifications

229°C to 231°C (decomposition)
Pink
C2H4CoO6
MFCD00050793
14,2445
MWHSMSAKVHVSAS-UHFFFAOYSA-L
cobalt(2+);oxalate;dihydrate
131881712
99.995%
Odorless
(metals basis)
5965-38-8
Powder
CoC2O4·2H2O
UN3288
cobaltousoxalate,cobaltoxalate
C(=O)(C(=O)[O-])[O-].O.O.[Co+2]
182.981
182.98 (146.95 Anhydrous)
Puratronic™
3.021 g/mL
Cobalt(II) oxalate dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c

H302+H312-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 212-409-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only