missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11375029
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Stabilizer for HCN, temperature indicator

Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.981
MDL Number MFCD00050793
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name cobalt(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Co+2]

Specifications

Melting Point 229°C to 231°C (decomposition)
Density 3.021 g/mL
Odor Odorless
Quantity 5 g
UN Number UN3288
Merck Index 14,2445
Solubility Information Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids.
Formula Weight 182.98 (146.95 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Cobalt (II) oxalate dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'