Learn More
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
49.55 € - 318.59 €
Chemical Identifiers
CAS | 1307-96-6 |
---|---|
Molecular Formula | CoO |
Molecular Weight (g/mol) | 74.93 |
MDL Number | MFCD00016031 |
InChI Key | IVMYJDGYRUAWML-UHFFFAOYSA-N |
Synonym | cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 |
PubChem CID | 14786 |
IUPAC Name | oxocobalt |
SMILES | O=[Co] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10177732
|
Thermo Scientific Acros
379200100 |
10 g | Glass bottle |
49.55 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15159273
|
Thermo Scientific Acros
379201000 |
100 g | Glass bottle |
318.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1307-96-6 | |
74.93 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
14786 | |
O=[Co] |
CoO | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
oxocobalt |
Specifications
1785.0°C | |
Brown-Green or Gray | |
99+% | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
oxocobalt | |
14786 | |
>74% (Co) |
1307-96-6 | |
Powder | |
CoO | |
15,243 | |
Solubility in water: insoluble | |
O=[Co] | |
74.93 | |
74.93 | |
Cobalt(II) oxide |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at rest in a posit
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-154-6
- Cobalt(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only