Learn More
Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis), Thermo Scientific Chemicals
125.00 € - 1650.00 €
Chemical Identifiers
CAS | 13478-33-6 |
---|---|
Molecular Formula | Cl2CoO8 |
Molecular Weight (g/mol) | 257.825 |
MDL Number | MFCD00150207 |
InChI Key | BSUSEPIPTZNHMN-UHFFFAOYSA-L |
Synonym | cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt |
PubChem CID | 26064 |
IUPAC Name | cobalt(2+);diperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438430
|
Thermo Scientific Alfa Aesar
044320.09 |
10 g |
125.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448430
|
Thermo Scientific Alfa Aesar
044320.18 |
50 g |
459.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458430
|
Thermo Scientific Alfa Aesar
044320.30 |
250 g | |||||||
Description
It is used as a chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13478-33-6 | |
257.825 | |
BSUSEPIPTZNHMN-UHFFFAOYSA-L | |
26064 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2] |
Cl2CoO8 | |
MFCD00150207 | |
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt | |
cobalt(2+);diperchlorate |
Specifications
13478-33-6 | |
Cl2CoO8 | |
UN1481 | |
Soluble in water. | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2] | |
257.825 | |
366.74 (257.83 Anhydrous) | |
Odorless | |
(metals basis) |
Crystalline | |
MFCD00150207 | |
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt | |
BSUSEPIPTZNHMN-UHFFFAOYSA-L | |
cobalt(2+);diperchlorate | |
26064 | |
99.999% | |
Hygroscopic | |
Cobalt (II) perchlorate hexahydrate |
Safety and Handling
GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P342+P311-P362-P370+P378q-P501c
H272-H315-H319-H334-H335-H350
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 236-653-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only