missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) selenide, 99+% (metals basis), Thermo Scientific Chemicals

224.12 € - 910.00 €

Chemical Identifiers

CAS 1307-99-9
Molecular Formula CoSe
Molecular Weight (g/mol) 137.904
MDL Number MFCD00016020
InChI Key QVYIMIJFGKEJDW-UHFFFAOYSA-N
Synonym cobalt selenide, cobalt ii selenide, cobalt selenide, cose, cobalt ii selenide trace metals basis 50g
PubChem CID 102108
IUPAC Name selanylidenecobalt
SMILES [Co]=[Se]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379377
View Documents
Thermo Scientific Alfa Aesar
040224.18
50 g
224.12 €
50g
Please to purchase this item. Need a web account? Register with us today!
11389377
View Documents
Thermo Scientific Alfa Aesar
040224.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1307-99-9
137.904
QVYIMIJFGKEJDW-UHFFFAOYSA-N
102108
[Co]=[Se]
CoSe
MFCD00016020
cobalt selenide, cobalt ii selenide, cobalt selenide, cose, cobalt ii selenide trace metals basis 50g
selanylidenecobalt

Specifications

1307-99-9
CoSe
UN3283
Soluble in HNO3,aqua regia
[Co]=[Se]
137.904
137.89
Weak
(metals basis)
Powder
MFCD00016020
cobalt selenide, cobalt ii selenide, cobalt selenide, cose, cobalt ii selenide trace metals basis 50g
QVYIMIJFGKEJDW-UHFFFAOYSA-N
selanylidenecobalt
102108
≥99%
7.650 g/mL
Cobalt(II) selenide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-155-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only