missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) sulfamate hydrate
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
Description
Cobalt(II) sulfamate hydrate is used in photonic device fabrication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Soluble in water. |
| IUPAC Name | λ²-cobalt(2+) disulfamate |
| Formula Weight | 251.11 (Anhydrous) |
| Odor | Odorless |
| Chemical Name or Material | Cobalt (II) sulfamate hydrate |
Sicherheit und Handhabung
Produktidentifikator
- Cobalt(II) sulphamate hydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Cobalt(II) sulphamate hydrate
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur