missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Sulfate, 98+%, Extra Pure, SLR, Fisher Chemical™

59.40 € - 97.45 €

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10763981
View Documents
Fisher Chemical
C/6840/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10773981
View Documents
Fisher Chemical
C/6840/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

10026-24-1
281.09
MEYVLGVRTYSQHI-UHFFFAOYSA-L
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
CoH14O11S
MFCD00149658
λ²-cobalt(2+) heptahydrate sulfate

Specifications

98°C
Brown-Red
Solid
MFCD00149658
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
281.09
Plastic powder jar
10026-24-1
4 to 5
CoH14O11S
MEYVLGVRTYSQHI-UHFFFAOYSA-L
λ²-cobalt(2+) heptahydrate sulfate
281.09g/mol
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.