missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) Sulfate Heptahydrate, 99-101%, Certified AR for Analysis, Fisher Chemical
Click to view available options
100g, Plastic powder jar
Specifications
Melting Point | 98°C |
CAS | 10026-24-1 |
Color | Brown-Red |
pH | 4 to 5 |
Physical Form | Solid |
Quantity | 100 g |
Molecular Formula | CoH14O11S |
MDL Number | MFCD00149658 |
InChI Key | MEYVLGVRTYSQHI-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Show More |
Safety and Handling
Product Identifier
- COBALTOUS SULFATE AR
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304-IF INHALED:.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : Cobalt(II) sulfate heptahydrate, Cobaltsulfate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'