missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) sulfate heptahydrate, Honeywell™
Click to view available options
Quantity:
100 g
250 g
1 kg
2.5 kg
Description
- ≥95% for general laboratory use.
Specifications
Quantity | 250 g |
Linear Formula | CoSO4 · 7H2O |
UN Number | UN3077 |
Formula Weight | 281.1g/mol |
Percent Purity | ≥99% |
Density | 2.03g/mL at 25°C |
Chemical Name or Material | Cobalt(II) sulfate heptahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'