missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11320959
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Color Red
pH 4
Physical Form Crystalline Aggregates
Quantity 100 g
Linear Formula CoSO4·7H2O
UN Number UN3077
Merck Index 14,2448
Solubility Information Soluble in water.
IUPAC Name λ²-cobalt(2+) heptahydrate sulfate
Formula Weight 281.10 (154.99 Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Density 2.03 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'