Learn More
Cobalt(II) sulfide, 99.5% (metals basis)
Description
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | >1116°C |
| Color | Red |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.5% (Metal basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 91 |
| Odor | Odorless |
| Appearance | Red powder |
| Density | 5.45g/cm3 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.