Learn More
Cobalt(II) tungsten oxide, 99%, Thermo Scientific Chemicals
pigment, drier for enamels, inks, and paints, electronic devices
65.70 € - 708.00 €
Chemical Identifiers
CAS | 10101-58-3 |
---|---|
Molecular Formula | CoO4W |
Molecular Weight (g/mol) | 306.77 |
MDL Number | MFCD00016034 |
InChI Key | BGLJOHZOBUGBBY-UHFFFAOYSA-N |
Synonym | cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten |
PubChem CID | 11738337 |
IUPAC Name | cobalt(2+);dioxido(dioxo)tungsten |
SMILES | [O--].[O--].[O--].[O--].[Co++].[W] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349377
|
Thermo Scientific Alfa Aesar
040223.09 |
10 g |
65.70 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369377
|
Thermo Scientific Alfa Aesar
040223.18 |
50 g |
167.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359377
|
Thermo Scientific Alfa Aesar
040223.30 |
250 g |
708.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pigment, drier for enamels, inks, and paints, electronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-58-3 | |
306.77 | |
BGLJOHZOBUGBBY-UHFFFAOYSA-N | |
11738337 | |
[O--].[O--].[O--].[O--].[Co++].[W] |
CoO4W | |
MFCD00016034 | |
cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten | |
cobalt(2+);dioxido(dioxo)tungsten |
Specifications
10101-58-3 | |
CoO4W | |
cobalt ii tungsten oxide, cobalt ii tungstate, co.wo4, cobalt 2+ ;dioxido dioxo tungsten | |
BGLJOHZOBUGBBY-UHFFFAOYSA-N | |
cobalt(2+);dioxido(dioxo)tungsten | |
11738337 | |
99% | |
Cobalt(II) tungsten oxide |
Powder | |
MFCD00016034 | |
Insoluble in water; Soluble in hot concentrated acids | |
[O--].[O--].[O--].[O--].[Co++].[W] | |
306.77 | |
306.78 | |
8.42 g/mL |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P201-P202-P281-P308+P313-P501c
H350
missing translation for 'einecsNumber' : 233-254-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only