Learn More
Cobalt(IV) sulfide, 99.5% (metals basis), Thermo Scientific Chemicals
130.00 €
Chemical Identifiers
CAS | 12013-10-4 |
---|---|
Molecular Formula | CoS2 |
Molecular Weight (g/mol) | 123.05 |
MDL Number | MFCD00064735 |
InChI Key | XUKVMZJGMBEQDE-UHFFFAOYSA-N |
Synonym | cobalt 3+ ;trisulfide, cobalt sulfide, |
PubChem CID | 25147475 |
SMILES | S=[Co]=S |
Description
Cobalt(IV) sulfide is a promising battery electrode material exhibiting good electrochemical properties for lithium sulfur batteries, thermal batteries and lithium ion secondary battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12013-10-4 | |
123.05 | |
XUKVMZJGMBEQDE-UHFFFAOYSA-N | |
25147475 |
CoS2 | |
MFCD00064735 | |
cobalt 3+ ;trisulfide, cobalt sulfide, | |
S=[Co]=S |
Specifications
12013-10-4 | |
Powder | |
MFCD00064735 | |
cobalt 3+ ;trisulfide, cobalt sulfide, | |
XUKVMZJGMBEQDE-UHFFFAOYSA-N | |
cobalt(3+);trisulfide | |
25147475 | |
99.5% | |
(metals basis excluding Ni), Ni <0.2% |
Black | |
CoS2 | |
UN3077 | |
Slightly soluble in acid. Insoluble in water. | |
S=[Co]=S | |
123.05 | |
123.06 | |
Odorless | |
Cobalt(IV) sulfide |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : GG3310000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only