missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Organometallic Standards, SPEX CertiPrep™

Product Code. 11995174 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Co1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-48-4, 64742-65-0
Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 99.9
Quantity 50 g
CAS Max % 0.1, 99.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Type Certified Reference Material
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Co
Solvent or Matrix Base Oil 75
MDL Number MFCD00010935
For Use With (Application) Petrochemical Testing
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Physical Form Solid
SMILES [Co]
IUPAC Name cobalt
Molecular Weight (g/mol) 58.93
PubChem CID 104730
ChEBI CHEBI:27638
Chemical Name or Material Cobalt Oil Standard
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1000 ug/g Cobalt in Base Oil 75
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : cobalt

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.