Learn More
Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045579.22
Description
Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina is used in the preparation of biofuel production by using algae.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt oxide, Molybdenum oxide | |
Blue | |
100 g | |
UN3077 | |
Co oxide:typically 3.4 to 4.5%; Mo oxide: typically 11.5 to 14.5% |
On alumina | |
Trilobe Extrudate | |
MFCD01074445 | |
Insoluble in water. | |
Odorless |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only