Learn More
Cobalt rod, 5mm (0.2in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
198.99 €
Chemical Identifiers
CAS | 7440-48-4 |
---|---|
Molecular Formula | Co |
Molecular Weight (g/mol) | 58.93 |
MDL Number | MFCD00010935 |
InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
PubChem CID | 104730 |
ChEBI | CHEBI:27638 |
IUPAC Name | cobalt |
SMILES | [Co] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334678
|
Thermo Scientific Alfa Aesar
043032.DJ |
50 mm |
198.99 €
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344678
|
Thermo Scientific Alfa Aesar
043032.DO |
150 mm |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-48-4 | |
58.93 | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
104730 | |
cobalt |
Co | |
MFCD00010935 | |
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
CHEBI:27638 | |
[Co] |
Specifications
7440-48-4 | |
MFCD00010935 | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
cobalt | |
104730 | |
99.95% | |
≈1.75g/cm | |
(metals basis) |
Co | |
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
[Co] | |
58.93 | |
CHEBI:27638 | |
Odorless | |
Rod | |
Cobalt rod, 5mm (0.2 in.) dia. |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350
missing translation for 'einecsNumber' : 231-158-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.