missing translation for 'onlineSavingsMsg'
Learn More

Melford Coelenterazine-H

Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA.

Brand:  Melford C61500-0.001

 View more versions of this product

Product Code. 30018190

  • 149.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA.

Exhibits 16-fold greater luminescence intensity as compared to native Coelenterazine.
  • CAS Number: 50909-86-9
  • Molecular Formula: C26H21N3O2
  • Molecular Weight: 407.5
Specifications

Specifications

50909-86-9
C26H21N3O2
1 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Melford Coelenterazine-H > 1 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.