missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Coelenterazine h

Coelenterazine h

Brand:  Molecular Probes™ C6780

218.12 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 11510166

  • 287.00 € / 250µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Coelenterazine h is one of several synthetic coelenterazine analogs that confer different Ca2+ affinities and spectral properties on the aequorin complex. Like native coelenterazine, these five novel derivatives can be used to reconstitute the aequorin complex both in vivo and in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Luminometer (Microplate), Luminometer (Tube-based)
Protein-Based
Room Temperature
Bioluminescence
Coelenterazine h
250 μg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.