Learn More
Melford Coelenterazine Native
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
Brand:  Melford C70900-0.01
 View more versions of this product
Product Code. 30018203
  • 453.00 € / 10mg
Explore more special offers
Description
Description
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion. Used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.
Specifications
Specifications
55779-48-1
C26H21N3O3