missing translation for 'onlineSavingsMsg'
Learn More

Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals

Product Code. 15438339
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 mg

A sensitive and specific chemiluminescence probe of the superoxide anion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55779-48-1
Molecular Formula C26H21N3O3
Molecular Weight (g/mol) 423.472
MDL Number MFCD00467176
InChI Key YHIPILPTUVMWQT-UHFFFAOYSA-N
Synonym coelenterazine, oplophorus luciferin, unii-3o1cb88rrd, clzn, coelenterate luciferin, coelenterazine, native, 3o1cb88rrd, 8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one, 2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one, 3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
PubChem CID 2830
ChEBI CHEBI:2311
IUPAC Name 8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
SMILES C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O

Specifications

Packaging Plastic bottle
Quantity 10 mg
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol or ethanol
Formula Weight 423.46
Physical Form Solid
Chemical Name or Material Coelenterazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.