missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Coenzyme Q10, 98%
52.66 € - 438.18 €
Chemical Identifiers
CAS | 303-98-0 |
---|---|
Molecular Formula | C59H90O4 |
Molecular Weight (g/mol) | 863.37 |
MDL Number | MFCD00042919 |
InChI Key | ACTIUHUUMQJHFO-UPTCCGCDSA-N |
Synonym | coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon |
PubChem CID | 5281915 |
ChEBI | CHEBI:46245 |
SMILES | COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15575276
|
Thermo Scientific Chemicals
457950010 |
1 g |
52.66 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15585276
|
Thermo Scientific Chemicals
457950050 |
5 g |
124.62 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15595276
|
Thermo Scientific Chemicals
457950250 |
25 g |
438.18 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
303-98-0 | |
863.37 | |
ACTIUHUUMQJHFO-UPTCCGCDSA-N | |
5281915 | |
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O |
C59H90O4 | |
MFCD00042919 | |
coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon | |
CHEBI:46245 |
Specifications
303-98-0 | |
Authentic | |
Glass Bottle | |
MFCD00042919 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and ethanol | |
COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O | |
863.37 | |
CHEBI:46245 | |
98% |
49.0°C | |
97.5% min. (HPLC) | |
C59H90O4 | |
coenzyme q10, ubidecarenone, ubiquinone 50, coq10, ubiquinone-10, neuquinon, ubiquinone, justquinon, neuquinone, emitolon | |
ACTIUHUUMQJHFO-UPTCCGCDSA-N | |
2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaenyl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione | |
5281915 | |
863.37 | |
Coenzyme Q10 |
Safety and Handling
missing translation for 'einecsNumber' : 206-147-9
RUO – Research Use Only