missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Cohesive Pocket Sampler

Specially adapted to enable the taking of samples of damp and cohesive powders. Sampling Systems™ Cohesive Pocket Sampler incorporates scrapers to push the sticky powder into the sample pockets.

Brand:  Sampling Systems™ 2220B-600

 View more versions of this product

Product Code. 15879311

  • 900.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured in 316L Stainless Steel
  • Multilevel discreet samples
  • Scrapers are fully welded to give a hygienic, GMP correct sampler
Specifications

Specifications

Sampler
3 Pockets
25 mm
Stainless Steel (316L)
3 x 15 mL
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sampling Systems™ Cohesive Pocket Sampler > LengthMetric: 600mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.