Learn More
Colchicine, 95% (dry wt.), Thermo Scientific Chemicals
78.30 € - 1275.00 €
Chemical Identifiers
CAS | 64-86-8 |
---|---|
Molecular Formula | C22H25NO6 |
Molecular Weight (g/mol) | 399.44 |
MDL Number | MFCD00078484,MFCD00078484 |
InChI Key | IAKHMKGGTNLKSZ-INIZCTEOSA-N |
Synonym | colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine |
PubChem CID | 45038708 |
SMILES | COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
11481920
|
Thermo Scientific Alfa Aesar
A13240.03 |
1 g |
78.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11491920
|
Thermo Scientific Alfa Aesar
A13240.06 |
5 g |
316.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401930
|
Thermo Scientific Alfa Aesar
A13240.14 |
25 g |
1275.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Colchicine is used as an anti-inflammatory agent for long-term treatment of behcet's Disease. It is used for constipation-predominant irritable bowel syndrome in women, and for treatment of severe or persistent aphthous stomatitis (canker sores).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water and alcohol. Sparingly soluble in ether.
Notes
Incompatible with strong oxidizing agents and mineral acids.
Identificatori chimici
64-86-8 | |
399.44 | |
IAKHMKGGTNLKSZ-INIZCTEOSA-N | |
45038708 |
C22H25NO6 | |
MFCD00078484,MFCD00078484 | |
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine | |
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 |
Specifica
64-86-8 | |
C22H25NO6 | |
UN1544 | |
Light sensitive | |
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine | |
IAKHMKGGTNLKSZ-INIZCTEOSA-N | |
2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide | |
45038708 | |
95% | |
Colchicine |
∼150°C (decomposition) | |
MFCD00078484,MFCD00078484 | |
2228813 | |
14,2471 | |
Soluble in water and alcohol. Sparingly soluble in ether. | |
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1 | |
399.44 | |
399.44 | |
(dry wt.) |
Sicurezza e movimentazione
GHS H Statement
H300-H340
Fatal if swallowed.
May cause genetic defects.
P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H300-H340
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 200-598-5
missing translation for 'rtecsNumber' : GH0700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only