Learn More
Unity Lab Services™ Cold Storage Equipment Requalification Service
Requalification Services for Cold Storage Equipment (documentation included)
Brand:  Unity Lab Services™ RQCold
Product Code. 30178681
  • 1950.00 € / 1 unit
Explore more special offers
Description
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cold Storage Equipment
Requalification Service