Learn More
VACUUBRAND™ Cold Trap
Use this Cold Trap with very volatile solvents and reduce the back streaming of oil molecules into the vacuum system. Vacubrand™ Cold Traps provide important protection for your pump by separating condensates and aggressive media.
1499.96 € - 2515.32 €
Specifications
For Use With (Equipment) Vacuubrand pumps
View More Specs
Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10428181
View Documents
VACUUBRAND™
20667051
SKF H 25 Cold Trap This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10548381
View accessory products View Documents
VACUUBRAND™
20667056
GKF 1000i Cold Trap This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10524902
View Documents
VACUUBRAND™
20667053
SKF H 40 Cold Trap This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Cold trap SKF
  • Sturdy, easy to clean
  • Easy to disassemble
  • Two-wall design with good conductance
  • Long operating time per coolant filling
  • Easy condensate drainage without disassembling
Cold trap GKF
  • Highly reflective insulation jacket for extended coolant life
  • Vertical window: Direct observation of condensate and coolant levels
  • PTFE stopcock: Drain condensate without disassembly
  • Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
  • Extended oil lifetime
  • Reduced service demands
  • Easy space-saving mounting
  • Simple filter change, service indicator
Specifications
Specifications
Vacuubrand pumps