Learn More
Lenz Laborglasintrumente™ Cold trap with Condensate Drain
Cold trap for direct filling of cooling agents
Brand:  Lenz Laborglasintrumente™ 05355546
 View more versions of this product
Product Code. 10420903
  • 566.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With valve stopcock (with retaining device)
  • For condensate drain
  • Low cooling agent consumption due to double-walled design
  • Apparatus connection: socket NS 29/32
  • Vacuum pump connection: cone NS 29/32
  • Made of DURAN™ tubing
Specifications
Specifications
DURAN Tubing
Valve stopcock (with retaining device) for condensate drain
200 mL
Cooling Agents
1