Learn More
Conductivity Solution, Oakton™
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
Brand:  Cole Parmer 0065315
 View more versions of this product
Product Code. 11765509
  • 18.20 € / Each
Explore more special offers
Description
Description
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Two-year shelf-life on all solutions
  • Conductivity-to-TDS Calibration Values: ppm NaCl: 737.1, ppm 442: 1050, ppm KCl: 757.1
Specifications
Specifications
Cole-Parmer Conductivity Standards
500 mL
Conductivity Standard
1.5 mS (1500 μS)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Statement
  • EUH210-Safety data sheet available on request.