missing translation for 'onlineSavingsMsg'
Learn More

Keck Standard Taper Clips

Ideal for glass and tubing assembly

Brand:  Keck 06840-00

157.70 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


Product Code. 15524443

  • 166.00 € / Pack of 24
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pack of 24 Assortment
  • Economical KECK adapters, clamps, and clips will not scratch glass or corrode
  • Color-coded for fast, easy size identification
  • Use quick disconnect adapters for safe, convenient tubing-to-tubing or tubing-to-glass connections
  • Use RAMP CLAMP tubing clamps for easy, one-handed tubing flow control
  • Select standard taper clips (in plastic or lacquer-coated steel) or ball-joint clips for secure glass-to glass connections
  • Economical standard taper clips securely hold conical joints under pressure without damaging glass
  • Polyoxymethylene (POM) acetal resin clips are safe to 302°F (150°C)
  • Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids)
Specifications

Specifications

24/Pk.
POM
Assorted
Taper Clamp
Ramp clamp tubing clamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Keck Standard Taper Clips > Keck Standard Taper Clips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.