Learn More
SCAT Collectors for HPLC
Collect several tubes into a single connector - no problem with the right collector! The tube connector is the most flexible connector of the SCAT system.
Brand:  SCAT 160141
41.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10064784
  • 52.55 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For a reliable and clean connection of hoses and capillaries for collecting liquids. SCAT collectors are available in versions with up to 8 connection options.The connections are either on the side or vertically, so that the tubes can be connected easily. Tubes with different diameters from 2.3 to 11.5 mm can be connected. The connectors are made of chemical-resistant PTFE, PFA or PP material.
Order Info
dimForUseWith
Specifications
Specifications
2-in-1 collector, vertical, PTFE/PFA, 2 x 2.3/3.2 mm OD
Tube Connector
2-in-1 Collector
PTFE
Fittings for 2 x 2.3/3.2 mm OD
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCAT Collectors for HPLC > 2-in-1 collector, vertical, PTFE/PFA, 2 x 2.3/3.2 mm OD