missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Library Quantification Kit

Invitrogen™ Collibri™ Library Quantification Kit includes a ready-to-use master mix optimized for Illumina NGS library quantification and a library dilution buffer

442.00 €

Specifications

Content And Storage Store at -20°C.
Detection Method SYBR
Format Kit
Sample Type DNA, RNA
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, QuantStudio™, ViiA™ 7 System, StepOne™ System, StepOnePlus™ System
View More Specs

Products 1
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15930523
View Documents
Invitrogen™
A38524500
500 Reactions
442.00 €
500 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Includes Platinum II Taq Hot Start DNA Polymerase and enables convenient and accurate quantification of NGS libraries
  • Contains all components needed for the amplification reaction, including primers and passive reference dye, and does not require any preparation before use
  • Due to the universal concentration of the passive reference dye in the master mix, the kit can be used with any qPCR instrument without adjustments
  • Accurate quantification of libraries made from various sample types, including degraded samples
  • Ease of use with all components ready to use -- no reagent preparation required
  • Visual cues - specially formulated dyes included to track pipetting steps
  • Platinum hot-start technology - superior specificity, sensitivity, and room-temperature reaction setup Unmatched convenience
  • Contains:
    • Platinum II Taq Hot Start DNA Polymerase
    • Optimized buffer
    • dNTPs
    • Primer mix targeting adapter sequences
    • SYBR green dye
    • Passive reference dye
    • Blue dye to track pipetting steps
  • Unique formulation provides ease of use at every step of the workflow:
    • No reagent preparation
    • Tracking of pipetting steps
    • One cycling protocol for libraries of various lengths and GC/AT-content levels
    • Universal passive reference dye concentration - a single master mix for all qPCR instruments
  • Also includes a ready-to-use library dilution buffer that contains stabilizing agents to enable library integrity after dilution. The buffer contains yellow dye to track pipetting steps. When combined, the blue master mix and yellow diluted libraries become a green solution. This feature eases tracking of the addition of the diluted libraries or DNA standards to the master mix in the qPCR plate wells, helping reduce pipetting errors.
Specifications

Specifications

Store at -20°C.
Kit
7500 Fast System, 7900HT Fast System, QuantStudio™, ViiA™ 7 System, StepOne™ System, StepOnePlus™ System
Illumina NGS Library Quantification
Platinum™ II Taq Hot Start DNA Polymerase
Library Quantification Kit
Standard
SYBR
DNA, RNA
Low
qPCR
Collibri™
Dry Ice
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.