missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ PCR-free PS DNA Library Prep Kits for Illumina Systems

The Invitrogen Collibri PCR-free PS DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes, including FFPE samples.

610.00 € - 2985.00 €

Specifications

Format Kit
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Application) Whole Genome Sequencing, Resequencing, De Novo Assembly
Libraries Fragment Library
View More Specs

Products 9
Product Code Brand No. of Reactions Product Type Price Quantity & Availability  
Product Code Brand No. of Reactions Product Type Price Quantity & Availability  
17707453
View Documents
Invitrogen™
A38608024W
24 Preps PCR-free PS DNA Library Prep Core Kit
610.00 €
Pack of 24
Please to purchase this item. Need a web account? Register with us today!
17717453
View Documents
Invitrogen™
A38610096W
96 Preps PCR-free PS DNA Library Prep Core Kit
2245.00 €
Pack of 96
Please to purchase this item. Need a web account? Register with us today!
16504311
View Documents
Invitrogen™
A38608024
24 Preps PCR-free PS DNA Library Prep Kit with CD Indexes
818.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16524311
View Documents
Invitrogen™
A38610096
96 Preps PCR-free PS DNA Library Prep Kit with CD Indexes
2985.00 €
96 reactions
Please to purchase this item. Need a web account? Register with us today!
16514311
View Documents
Invitrogen™
A38609024
24 Preps PCR-free PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
890.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16564321
View Documents
Invitrogen™
A43608024
24 Preps PCR-free PS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
890.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16584321
View Documents
Invitrogen™
A43609024
24 Preps PCR-free PS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
890.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
16504331
View Documents
Invitrogen™
A43610024
24 Preps PCR-free PS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
890.00 €
24 reactions
Please to purchase this item. Need a web account? Register with us today!
17143149
View Documents
Invitrogen™
A38615196
96 Preps PCR-free PS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
2985.00 €
Pack of 96
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Collibri PCR-free PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from inputs as low as 500 ng of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PCR-free PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from inputs as low as 500 ng of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PCR-free PS DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri PCR-free PS DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. The kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification and sequencing on Illumina instruments.

Applications
The Collibri PCR-free PS DNA Library Preparation Kit is recommended for:
Whole genome sequencing
• Resequencing
• De novo assembly

Specifications

Specifications

Kit
Genome and DNA Sequencing
Fragment Library
Dry Ice
DNA
Whole Genome Sequencing, Resequencing, De Novo Assembly
Collibri™
Library Generation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.