Learn More
Fisherbrand™ Colony Counter Bundle
Quickly identify and tally colonies with precision using this all-inclusive Colony Counter Bundle.
Brand:  Fisherbrand™ 120951
Product Code. 17842966
  • 595.00 € / 1 set
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quickly identify and tally colonies with precision using this all-inclusive package designed to streamline all colony counting procedures.
  • Fisherbrand eCount Colony Counter: Combines an electronic counter with a Sharpie™ pen for marking to prevent missing or double-counting colonies. Features count verification with beeps or LED flashes, dual counting modes (up and down), a display for total counts (up to 32 separate counts), and the ability to disable the counter for writing totals and notes.
  • Fisherbrand Lightbox: The Fisherbrand Lightbox serves as a LED light source for efficient bacterial colony counting. It features a spacious viewing area (125 x 100 mm) suitable for various samples, and is powered by six 1.5V AAA batteries (included).
  • Fisherbrand Magnifier: The Fisherbrand Mini-Magnifier enhances object visibility by providing 1.75X magnification. It features a swiveling, molded lens for comfortable observation.
  • Fisherbrand Silicone Lab Mat: This FDA food-grade silicone labmat is suitable for various laboratory settings, including cold rooms, clean rooms, and sterile suites, where high sterility is essential. Effectively safeguards countertops from temperatures up to 200°C (392°F) and demonstrates general chemical resistance to moderate oxidizing chemicals.
Specifications
Specifications
Colony Counter with Lightbox, Magnifier and Lab Mat
Digital
Up to 999 colonies per zones, Up to 32 individual zones
3 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Colony Counter Bundle > Fisherbrand Colony Counter