missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Coded pH Buffer Solution Tested at 20°C, Reagecon™
Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.
Brand: Reagecon 1070C
This item is not returnable.
View return policy
Description
- Colored pH Buffer Solutions
- Tested at 20°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
Specifications
0°C | |
7.00 | |
1 L |
Yellow | |
Liquid | |
Characteristic |