Learn More
Cytiva Adapter for XK Columns
Movable adapter gives a uniform flow that maintains the integrity of the packed bed during HPLC operations. Cytiva Adapter for XK Columns is an assembled unit consisting of a top-end cap, a plunger and an adjusting knob. Available for 16, 26 and 50mm diameter XK columns.
325.08 € - 494.00 €
Specifications
For Use With (Application) Liquid Chromatography at Low to Medium Pressure
Includes Plunger, Top-end cap, Adjusting knob
Quantity 1 Pack
View More Specs
Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11320352
View Documents
Cytiva
28-9898-76
XK Empty Column, 16 mm Diameter
325.08 €
Each
In Stock
to see stock.
Add to basket
11330352
View Documents
Cytiva
28-9898-77
XK Empty Column, 26 mm Diameter
415.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11340352
View Documents
Cytiva
28-9898-80
XK Empty Column, 50 mm Diameter
494.00 €
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
XK empty columns are designed for liquid chromatography at low to medium pressure. They are user friendly, robust, and give reproducible packing, and they feature fittings for direct connection to ÄKTA™ design instruments or other HPLC systems. XK columns are supplied with a movable adapter.
  • Plunger design ensures a homogeneous buffer distribution and reproducible packing
  • QuickLock mechanism of the adapter shaft facilitates easy column handling and cleaning
  • Adapter top-end cap fits directly into the column end piece
  • Plunger carries the net ring and support screen for the medium bed
  • An adjusting knob seals the adapter to the column wall
Specifications
Specifications
Liquid Chromatography at Low to Medium Pressure
Plunger, Top-end cap, Adjusting knob
1 Pack