Learn More
Lenz Laborglasinstrumente™ Column Head with Valve Stopcock
Made of DURAN™ tubing. Lenz Laborglasinstrumente™ Column Head with Valve Stopcock has a PTFE for easy and accurate adjustment and ground joint NS 14/23 for thermometers with a fitting length of 55 mm.
Brand:  Lenz Laborglasinstrumente™ 05170111
Product Code. 10274481
  • 1180.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With PTFE valve
  • Made of DURAN™ tubing
Specifications
Specifications
NS 14/23
DURAN Tubing
PTFE Valve
1 Each