missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Comacier VHP Plus Heavy Duty Leather Gloves

Product Code. 15565746
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Provides protection against cuts with flexibility and comfort. Ansell Comacier VHP Plus Heavy Duty Leather Gloves are made from high performance fibres using Techcor™ Knitting Technology.

  • Mechanical protection heavy duty gloves
  • Medium weight 10 gauge seamless knitted glove
  • Knit wrist ensures good fit
  • Unique traceability system on textile tag
  • Palm, fingers and artery guard made of leather for optimal protection
  • Provides the highest levels of mechanical resistance
  • Good flexibility and comfort
  • Superior grip when used with oils and greases due to leather patch

Applications:

  • Handling heavy and oily metal pieces
  • Handling sharp or irregular-edged metal parts
  • Press and stamping
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Color Gray
Exterior Finish Leather Palm
Material Leather
Product Type Mechanical Protection Gloves
Size 9
Glove Cuff Style Knit
Liner Material Techcor™
Packaging Type 12 pairs in bag, 10 bags in carton
Series VHP Plus
Length (Metric) 240 to 260 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'