missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Multi-Spin PVC 6000 Centrifuge/Vortex Mixer

Highly versatile and efficient variable-speed combined centrifuge/vortex mixer. Programmed centrifugation and mix operations or independent centrifuging and vortex-mixing of multiple microtubes and 0.2ml strips.

Brand:  Grant Instruments™ PCV-6000

704.82 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.


Product Code. 15261166

  • 759.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Spin-mix-spin technology can save considerable time by automatically performing a cycling program of sample mixing and spinning 12 tubes at once, when compared with removing the tubes for vortexing after every spin.
  • Temperature setting range +4ºC to +40ºC
  • Centrifugation speed: fixed at 2800rpm (50Hz) and 3500rpm (60Hz)
  • Choice of interchangeable rotors for different microtube sizes/combinations and for 0.2mL strips
  • Continuous operation or short spin
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000 to 6000 rpm
1.5/2 mL, 0.5 mL, 0.2 mL
235 mm
12 x 1.5/2.0 mL/12 x 0.5 mL plus 12 x 0.2 mL
120/230 V
125 mm
190 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.