missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combined Ion-Selective Electrode

Converts activity of a specific ion dissolved in a solution into an electrical potential. Fisherbrand™ Combined Ion-Selective Electrode is compatible with all pH/mV meters or ionometers.

504.56 € - 593.60 €

Specifications

Filling Solution Type ISAB/ISE
Electrode Combined ion-selective
View More Specs

Products 4
Product Code Brand Applications Interferences pH Range Price Quantity & Availability  
Product Code Brand Applications Interferences pH Range Price Quantity & Availability  
11795638
View accessory products View Documents
Fisherbrand™
NH41522-S7
Determination of Ammonium (NH4+) ion K+ 4 - 10
593.60 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11779708
View accessory products View Documents
Fisherbrand™
CU01522-S7
Determination of Copper (Cu2+) ion Ag+, Hg2+, Cl-, Br-, Cd2+, Fe2+ 2 - 12
518.00 €
Each
Estimated Shipment: 03-03-2025
to see stock.
Add to basket
 
11769708
View accessory products View Documents
Fisherbrand™
CL01522-S7
Determination of Chlorine (Cl-) ion S2-, I-, CN-, Br- 2 - 12 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11749718
View accessory products View Documents
Fisherbrand™
NO31522-S7
Determination of NO3- ion CIO4-, I-, CN-, BF4-, Cl-, NO2- 2.5 - 11
590.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • With solid membrane Interchangeable flat surface
  • Ion to be detected passes through the membrane generating a difference of potential, proportional to ionic concentration
  • Analysis time is generally less than one minute
  • Sample preparation steps such as filtering or Distillation rarely necessary
  • Calibration should be carried out as often as possible (once a day) at the same temperature as the sample
  • Measurement temperature 0 to 50°C
  • Delivered with membrane, electrolytic solution, ISAB solution and ISE standard and gasket
ISAB solution
: To adjust the ionic strength. add directly to the solution to be measured.
ISE standard
: For the calibration of ionic electrodes. Concentration 1000 ppm.
Specifications

Specifications

ISAB/ISE
Combined ion-selective
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.