Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
22.00 € - 30.90 €
Specifications
Type Comb
Quantity 1 Each
Thickness (Metric) 0.75 mm
View More Specs
Products 9
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Price Quantity & Availability  
11883303
View Documents
Fisherbrand™
MS10-1-0.75
1 Prep, 1 Marker 270 μL
22.00 €
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
11893303
View Documents
Fisherbrand™
MS10-10MC-0.75
10 20 μL
22.00 €
Each
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
11863313
View Documents
Fisherbrand™
MS10-12-0.75
12 17 μL
30.90 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
11803323
View Documents
Fisherbrand™
MS10-16-0.75
16 12 μL
23.90 €
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11843323
View Documents
Fisherbrand™
MS10-2-0.75
2 Prep, 2 Marker 57 μL
22.00 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
11853323
View Documents
Fisherbrand™
MS10-20-0.75
20 10 μL
22.00 €
Each
In Stock
to see stock.
Add to basket
11823333
View Documents
Fisherbrand™
MS10-25-0.75
25 7 μL
27.30 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
11863333
View Documents
Fisherbrand™
MS10-4-0.75
4 Prep, 2 Marker 118 μL
22.00 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
11803343
View Documents
Fisherbrand™
MS10-8-0.75
8 30 μL
22.25 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
  • For tightly resolved bands
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
1 Each
0.75 mm