Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ MS15-4-1
 View more versions of this product
Product Code. 11813413
  • 27.05 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Supplied as standard
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
4 Prep, 2 Marker
122 μL
Comb
1 mm