Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
19.60 € - 27.80 €
Specifications
For Use With (Equipment) OmniPAGE Mini Vertical Gel Electrophoresis Unit
Type Comb
Quantity 1 Each
Thickness (Metric) 1 mm
View More Specs
Products 8
Product Code Brand No. of Wells Volume (Metric) Well Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Well Price Quantity & Availability  
11877583
View Documents
Fisherbrand™
VS10-10-1
10 40 μL
19.60 €
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
11807593
View Documents
Fisherbrand™
VS10-1-1
1 Prep, 1 Marker 650 μL
20.10 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
11857593
View Documents
Fisherbrand™
VS10-12-1
12 35 μL
27.80 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
11807603
View Documents
Fisherbrand™
VS10-16MC-1
16 25 μL
27.20 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
11857603
View Documents
Fisherbrand™
VS10-20-1
20 20 μL
19.70 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
11807613
View Documents
Fisherbrand™
VS10-5-1
5 100 μL
19.70 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
11857613
View Documents
Fisherbrand™
VS10-8MC-1
8 60 μL
20.95 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
11807623
View Documents
Fisherbrand™
VS10-9-1
9 50 μL
19.70 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
Description
Description
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Supplied as standard
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
OmniPAGE Mini Vertical Gel Electrophoresis Unit
Comb
1 Each
1 mm