Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™ VS10-20-2
 View more versions of this product
Product Code. 11877603
  • 21.25 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Blue
OmniPAGE Mini Vertical Gel Electrophoresis Unit
20
2 mm
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
Comb
1 Each
40 μL