missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit

For use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis Unit

80.50 € - 83.75 €

Specifications

For Use With (Equipment) TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit
Type Comb
View More Specs

Products 10
Product Code Brand No. of Wells Volume (Metric) Sample Width (Metric) Teeth Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Sample Width (Metric) Teeth Price Quantity & Availability  
11736185
View Documents Promotion Details
Fisherbrand™
TVGC1512
12 35 μL 5.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11796185
View Documents Promotion Details
Fisherbrand™
TVGC1520
20 15 μL 2.5 mm
80.50 €
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
11726185
View Documents Promotion Details
Fisherbrand™
TVGC158
8 65 μL 9.25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11716175
View Documents Promotion Details
Fisherbrand™
TVGC18
8 45 μL 9.25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11706195
View Documents Promotion Details
Fisherbrand™
TVGC28
8 90 μL 9.25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11706185
View Documents Promotion Details
Fisherbrand™
TVGC112
12 25 μL 5.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11724925
View Documents Promotion Details
Fisherbrand™
TVGC115
15 20 μL 4 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11716185
View Documents Promotion Details
Fisherbrand™
TVGC120
20 10 μL 2.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11716195
View Documents Promotion Details
Fisherbrand™
TVGC212
12 50 μL 5.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11726195
View Documents Promotion Details
Fisherbrand™
TVGC220
20 20 μL 2.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Available in 1, 1.5 and 2mm thickness
Specifications

Specifications

TVG-SYS Vari-Gel MIDI Horizontal Gel Electrophoresis Unit
Comb
Videos
SDS
Documents

Documents

Product Certifications