Learn More
R&D Systems™ CometAssay Lysis Solution (2 x 500 mL)
For use with Single Cell Gel Electrophoresis Assay
Brand:  R&D Systems™ 4250-050-01
Product Code. 30002113
Description
Description
Cells are lysed using Lysis solution. Cool to 4°C, or on ice, for at least 20 minutes before use.
Specifications
Specifications
2 x 500 mL
CometAssay Lysis Solution