missing translation for 'onlineSavingsMsg'
Learn More

3M™ Forehead Comfort Bands for use with 3M™ Versaflo™ S-Series Headcovers and Hoods

Product Code. p-8006561
Click to view available options
Size:
Small
Large
Model:
S-957L
S-957S
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 15729027

Brand: 3M™ 7000044309

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to create a more comfortable fit for head covers and hoods, the 3M™ Forehead Comfort Band attaches to the head suspension headband to cushion the user's forehead. Available in various sizes.

  • Product Type: Forehead Comfort Band
  • Description: Standard Format, CE-Approved
  • Color: Grey
  • For Use With (Equipment): 3M™ Versaflo™ S-Series Headcovers and Hoods
  • For Use With (Application): Respiratory Protection
TRUSTED_SUSTAINABILITY

Specifications

Description Standard, CE-Approved
Color Gray
Disposable Reusable
Product Type Comfort Band
Size Small
For Use With (Application) Respiratory Protection
For Use With (Equipment) Versaflo S-Series Headcovers and Hoods
Model S-957S
Quantity 20 Pk.
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.