Learn More
Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand:  Tenak™ TE24292
 View more versions of this product
Product Code. 12435830
  • 175.00 € / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
Specifications
Specifications
100 mm (Box)
Boxes (4)
Stainless Steel
Upright Freezers
2 x 2